Chart Recorder for Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
3166207
13306-252EA 3130 USD
13306-252
Chart Recorder for Incubators, Thermo Scientific
Chart Recorders Recorders
This circular digital chart recorder is designed to use with the automatic water jacketed CO2 incubator, model 815 low-temperature incubators and model 818 diurnal incubators.