Petri Dish Storage Canisters, Boekel Scientific
Supplier: Boekel
1370S
13278-220EA 52.04 USD
13278-220 13278-216
Petri Dish Storage Canisters, Boekel Scientific
Racks Petri Dish Racks
Designed to store Petri Dishes, these canisters are ideal for organizing and sterilizing glass Petri Dishes
These stainless steel canisters keep dishes stacked and within reach.