VWR® Bath Fluids
Supplier: VWR International
These heat-transfer fluids provide superior thermal stability for high or low temperatures.
- Silicone heat transfer fluid
Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.
Packaging: Bottles are packaged in a carton for shipment.