VWR® Modular Heating Block for Titer Plates
Supplier: VWR International
13259-295
13259-295EA 454.86 USD
13259-295
VWR® Modular Heating Block for Titer Plates
Block Heaters Blocks
Heating block is ideal for use with 96-well or 384-well titer plates.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
It features a recessed well for better stability and a flat surface appropriate for flat or round-bottom plates. Double block size fits only 2, 4, or 6 block heaters.