SP Bel-Art Inhalation Narcosis Chamber, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H46420-0002
13197-460EA 704.46 USD
13197-460
SP Bel-Art Inhalation Narcosis Chamber, Bel-Art Products, a part of SP
Animal Supplies Animal Anesthesia Systems
A clear acrylic, hinged cover permits observation of small animals during anesthetizing.
  • Clear acrylic cover
  • Bottom chamber contains absorptive cotton
  • Folding perforated platform
  • Polypropylene components
  • Unaffected by anesthetic compounds
Cover and additional polypropylene components are not affected by anesthetic compounds
A stopcock permits entrance of fresh air during anesthesia to prevent suffocation and accepts 4 mm-6 mm ID tubing. Maximum I.D. is 19.7 cm (7¾”), top I.D. is 17.8 cm (7”), height from platform to top is 12.7 cm (5”), height under platform is 5.7 cm (2¼”), overall height is 19.7 cm (7¾”).
A folding perforated platform is removed to fill the bottom with cotton and replaced to support the animal. Anesthetic is poured into the small funnel at the side and runs into the bottom of the chamber where it is absorbed by cotton.