SP Bel-Art® Bellows Samplers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37823-0050
13197-432CS 431.37 USD
13197-432
SP Bel-Art® Bellows Samplers, Bel-Art Products, a part of SP
Samplers Fluid Samplers
Bellows samplers provide quick and easy liquid sampling with one-handed operation.
  • Polyethylene construction
  • Long, tapered tip
  • Quick and easy sampling
  • One-handed operation
The FDA-grade polyethylene sampler tube has a long tapered tip with a 4 mm opening.