SP Bel-Art Stirring Paddle, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37770-0000
13197-418EA 165.54 USD
13197-418
SP Bel-Art Stirring Paddle, Bel-Art Products, a part of SP
Stirrers Stirring Tools
Designed for mixing liquids in large vats and tanks, this high-density polyethylene stirring paddle has a 91 cm (3') handle.
  • HDPE construction
  • Designed for use with large vats and tanks
  • Long handle extends reach and provides extra safety
The paddle measures 7.6x15.2 cm (3x6"), and the handle has a 25 mm (1") diameter.