SP Bel-Art Teflon® Policeman, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37738-0000
13197-416CS 398.14 USD
13197-416
SP Bel-Art Teflon® Policeman, Bel-Art Products, a part of SP
Spatulas
Made of a 7.9 mm (⁵/₁₆) Ø Teflon® PTFE rod with flattened ends, this policeman is useful for scraping and transferring materials.
  • Useful for scraping and transferring materials
  • Durable PTFE construction
  • Flattened ends
  • Autoclavable
Steam autoclavable at 121 °C (250 °F).