SP Bel-Art Resharpenable Rigid Scraper, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36820-0000
13197-404EA 100.37 USD
13197-404
SP Bel-Art Resharpenable Rigid Scraper, Bel-Art Products, a part of SP
Scrapers
Made from glass-reinforced polyimide nylon, this rigid scraper's straight, chisel-type edge may be resharpened.
  • Glass-reinforced polyimide nylon construction
  • Rigid design
  • Straight edge
  • Wide, flat chisel-type blade
It has a 14 cm (5½”) long x 6.3 cm (2.5”) wide flat blade. FDA-grade materials are suitable for direct contact with foods, drugs, and cosmetics. Highest temp: 180ºF (82ºC).