SP Bel-Art Coated Lead Shield, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H24958-0000
13197-364EA 630.56 USD
13197-364
SP Bel-Art Coated Lead Shield, Bel-Art Products, a part of SP
Shields Radiation Shields
This 6.4 mm (¼") thick lead shield safeguards against chemical reactions, low-level radiation, and flying debris.
  • Vinyl coating
  • Thick lead construction
  • Protects against chemicals, low-level radiation, and debris
The lead shield is coated with vinyl for a soft, smooth surface that prevents the hazard of skin contact with lead. Weight 3.6 kg (8 lbs). Other sizes made to order.