SP Bel-Art Vacuum Chamber and Plate, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42043-0000
13197-270EA 313.45 USD
13197-270
SP Bel-Art Vacuum Chamber and Plate, Bel-Art Products, a part of SP
Desiccators Desiccators, Plastic
This vacuum chamber is useful for vacuum drying, degassing, and teaching demonstrations.
  • Polypropylene stopcock construction
  • TFE resin plug
  • Clear polycarbonate vacuum chamber construction
  • ABS plastic plate
The chamber top is molded of clear polycarbonate for full visibility and will withstand a full vacuum at room temperature. A strong ABS plastic plate with an O-ring seals against the chamber’s flange. The polypropylene stopcock with Teflon® TFE resin plug accepts 6.4 mm (1/4") I.D. tubing.