SP Bel-Art Cup-Type Stoppers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H20193-0002
13195-190PK 72.55 USD
13195-190 13196-088 16126-498 30617-646
SP Bel-Art Cup-Type Stoppers, Bel-Art Products, a part of SP
Stoppers PE/LDPE/HDPE Stoppers
These polyethylene cup-type stoppers fit tightly yet remove easily.
  • Polyethylene construction
  • Tight fit, yet easy to remove
  • Cup style allows them to double as micro containers
They also double as containers for micro work. All are 26 mm high.