VWR® Talon® Support Stand, Cast Iron, Tripod Base
Supplier: VWR International
12985-076
12985-076EA 150.93 USD
12985-076
VWR® Talon® Support Stand, Cast Iron, Tripod Base
Support Stands
Support stand base is constructed of cast iron with black enamel finish.
  • Cast iron construction with black enamel finish
  • Tripod base
  • Built-in support rod holder with locking knob
  • For use with VWR® Talon® lab frames
Base features a built-in support rod holder with locking knob that accepts a 1.3 cm (1/2") diameter support rod (sold separately).