VWR® Traceable® Infrared Thermometer Gun
Supplier: VWR International
12777-846
12777-846EA 283.1 USD
12777-846
VWR® Traceable® Infrared Thermometer Gun
Thermometers Digital Thermometers
Small, lightweight unit takes temperature readings of any surface in less than one second.
  • Traceable®
  • Lightweight
  • Fast temperature readings
  • 11:1 field of view
Laser sighting beam permits direct aim at target surface
The emissivity rate is fixed at 0.95. The field of view is 11:1, indicating that the gun can read a spot the size of 5 cm (2") from 55.9 cm (22") away.
Features include backlighting, data hold (freezes display reading), laser, low-battery indicator, and a °C/°F button.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with 9V battery.