VWR® Pocket Dew Point/Hygrometer/Thermometer
Supplier: VWR International
12777-834 10754-550 10754-732
12777-834EA 241.82 USD
12777-834 10754-550 10754-732
VWR® Pocket Dew Point/Hygrometer/Thermometer
Hygrometers Digital Thermo-Hygrometers
Pocket-size meter registers relative humidity with either dew point or temperature on a 1.9 cm (3/4") high dual-display LCD.
  • Traceable®
  • Compact design
The device is fitted with a rugged ABS plastic case and an electronic capacitance polymer film sensor that is unaffected by condensation. At the press of a button, the unit recalls the minimum/maximum of all readings; the hold button “freezes” the display to record a reading. Typical full-range response time is a fast 30 to 60 seconds.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two AAA batteries.