Lockout for Miniature SMC Models AR2000/NAR2000, Brady Worldwide®
Supplier: Brady Worldwide
64539
12222-826EA 64.39 USD
12222-826
Lockout for Miniature SMC Models AR2000/NAR2000, Brady Worldwide®
Security Locks
Lockout enables regulator to be locked in the on, off, or throttled position
Easy installation involves removing snap cap, installing lockout on regulator rod, and snapping replacement cap on regulator. Supplied with lockout, cap, and allen wrench.