Hubbell Plugout™, Brady Worldwide®
Supplier: Brady Worldwide
65695 65968
12222-750EA 80.82 USD
12222-750 12222-748
Hubbell Plugout™, Brady Worldwide®
Security Locks
Fits the majority of heavier industrial plug applications
The smaller version accepts plugs with a maximum 70mm (2.75") diameter and 12cm (4.75") length with 25mm (1") cord. The larger version accepts plugs with a maximum 114.3mm (4.5") diameter and 25.8cm (10") length with 54mm (2") cord.