Portable Metal Lock Box, Brady®
Supplier: Brady Worldwide
65699
12222-698EA 234.86 USD
12222-698
Portable Metal Lock Box, Brady®
Security Locks
Made of heavy-duty steel and powder-coated for extra rust resistance
  • Made of heavy-duty steel and powder coated for extra rust-resistance
  • Requires fewer locks and is lighter than hasps on delicate energy isolating devices like circuit breakers
  • Accommodates up to 13 padlocks (locks not included), can be used as lock storage box for up to 40 padlocks
  • Front slot allows keys to be inserted when box is locked
Requires fewer locks and is lighter than hasps on delicate energy isolating devices like circuit breakers. Accommodates up to 13 padlocks (locks not included) and can also be used as a lock storage box holding up to 40 padlocks. Front slot allows keys to be inserted when box is locked.