Long Ranger™ Sequencing Reagents, Lonza

Supplier: LONZA WALKERSVILLE INC

50611 50640
12001-750EA 542.47 USD
12001-750 12001-752 12001-812
Long Ranger™ Sequencing Reagents, Lonza
Nucleic Acid Reagents Sanger Sequencing Reagents

Long Ranger™ sequencing reagents are designed for the high performance sequencing on ABI Prism® 377 and Li-COR® Sequencers.


  • Long Ranger™ gel solution offers expceptional results for sequencing and STR analysis
  • Applications: DNA and RNA sequencing, STR analysing, identify testing, genotyping, different display
  • Storage temperature upto 18 - 26 °C


Single™ pack contains all necessary reagent for casting a single gel in a pre-measures, ready to use pouch. Gel™ Slick solution is a safe alternative to hazardous silance based coating solution and it is suitable for sequencing gel and coating microcentrifuge tubes.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR