SYBR® Green II nucleic acid stain

Supplier: LONZA WALKERSVILLE INC

50523
12001-802EA 737.55 USD
12001-802
SYBR® Green II nucleic acid stain
SYBR® Green II

These stains are fluorescent dyes used to detect DNA and RNA.


  • Sensitive
  • Less mutagenic
  • Less signal to noise ratio
  • Nucleic Acid Gel Staining


SYBR® Green stains are fluorescent dyes used to detect DNA and RNA. They are more sensitive and less mutagenic than standard stains, yet they provide excellent signal-to-noise ratio with minimal background. SYBR® Green I detects as little as 60 pg dsDNA and 1 ng oligonucleotides in agarose and polyacrylamide gels. It is optimal for PCR, apoptosis studies, and heteroduplex analysis.
SYBR® Green II detects 100 pg ssDNA and 2 ng RNA in agarose and polyacrylamide gels. SYBR® Green II is optimal for RNA gel electrophoresis and SSCP analysis. Both stains are supplied as a 10000X concentrated solution in DMSO.

MDL Number: MFCD00284716
CAS Number: 172827-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR