SYBR® Green I nucleic acid stain

Supplier: LONZA WALKERSVILLE INC

50512 50513
12001-796EA 1321.68 USD
12001-796 12001-798
SYBR® Green I nucleic acid stain
SYBR® Green I

These stains are fluorescent dyes used to detect DNA and RNA.


  • Sensitive
  • Less mutagenic
  • Less signal to noise ratio
  • Nucleic Acid Gel Staining


SYBR® Green stains are fluorescent dyes used to detect DNA and RNA. They are more sensitive and less mutagenic than standard stains, yet they provide excellent signal-to-noise ratio with minimal background. SYBR® Green I detects as little as 60 pg dsDNA and 1 ng oligonucleotides in agarose and polyacrylamide gels. It is optimal for PCR, apoptosis studies, and heteroduplex analysis.
SYBR® Green I is suitable for gel-based detection of small amounts of DNA in low copy/target number PCR products, apoptosis studies, and heteroduplex analysis. It detects 60 pg dsDNA and 1 ng oligonucleotide. It is also ideal for non-isotopic detection of DNA mutations by single-stranded conformation polymorphism (SSCP) analysis. It detects 100 pg of ssDNA and 2 ng RNA.


SYBR is a registered trademark of Thermo Fisher Scientific.

Formula: C₃₂H₃₇N₄S
MDL Number: MFCD00284715
CAS Number: 163795-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR