AccuGENE® 1X TE Buffers
Supplier: LONZA WALKERSVILLE INC
51235
12001-412EA 121.37 USD
12001-412
AccuGENE® 1X TE Buffers
Buffers Cloning Buffers
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs
  • Composition - 10 mM Tris, 1 mM EDTA (disodium salt), pH 7.4
TE buffer has traditionally been used to solubilize DNA or RNA, while protecting it from degradation. A common buffer, 'TE' is derived from its components: Tris, a common pH buffer, and EDTA, a molecule that chelates cations like Mg2+. Lonza’s AccugeneTM buffer is the perfect choice for DNA/RNA storage or any of the many other applications TE buffer is used for.
Store all reagents at 18 to 26 °C (64 to 79 °F).
18 megOhm water is filtered using a 0.2 µm filter, and filled into sterile bottles.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccb68259a9c3e576f1cc1743221ae6a0443c578e&vid=294849e634a4f80b3ffb44a5419b67fdcebf1ee8&prid=web&clid=SAPDAM