Anti-CD262 (DR5, TRAIL-R2) Armenian Hamster Monoclonal Antibody (Biotin) [clone: MD5-1]

Supplier: Biolegend
119904
119904-BLEA 462.6 USD
119904-BL
Anti-CD262 (DR5, TRAIL-R2) Armenian Hamster Monoclonal Antibody (Biotin) [clone: MD5-1]
Antibodies
Biotin anti-mouse CD262 (DR5, TRAIL-R2) [MD5-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD262 (DR5, TRAIL-R2)
Clonality: Monoclonal
Clone: MD5-1
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR