Anti-MD-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MD-113]

Supplier: Biolegend
117805
117805-BLEA 240.8 USD
117805-BL
Anti-MD-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MD-113]
Antibodies
PE anti-mouse MD-1 [MD-113]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: MD-1
Clonality: Monoclonal
Clone: MD-113
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR