Anti-H-2Kk Mouse Monoclonal Antibody (Biotin) [clone: 36-7-5]

Supplier: Biolegend
114903
114903-BLEA 93.4 USD
114903-BL
Anti-H-2Kk Mouse Monoclonal Antibody (Biotin) [clone: 36-7-5]
Antibodies
Biotin anti-mouse H-2Kk [36-7-5]; Isotype: Mouse (A.TL) IgG2a, κ; Reactivity: Mouse k haplotype, Weakly cross-reacts with SJL/Hsd mice splenocytes, but does not react with alloantigens of other haplotypes (e.g., b, d, q).; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kk
Clonality: Monoclonal
Clone: 36-7-5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse (A.TL) IgG2a, κ
Reactivity: Mouse k haplotype;Weakly cross-reacts with SJL/Hsd mice splenocytes;but does not react with alloantigens of other haplotypes (e.g.;b;d;q).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR