Anti-H-2Kb/H-2Db Mouse Monoclonal Antibody [clone: 28-8-6]

Supplier: Biolegend
114602
114602-BLEA 296.4 USD
114602-BL
Anti-H-2Kb/H-2Db Mouse Monoclonal Antibody [clone: 28-8-6]
Antibodies
Purified anti-mouse H-2Kb/H-2Db [28-8-6]; Isotype: Mouse (C3H) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with H-2Dd MHC class I alloantigen. Does not react with other haplotypes (e.g., f, k, p, q, r, s); Apps: FC, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb/H-2Db
Clonality: Monoclonal
Clone: 28-8-6
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse (C3H) IgG2a, κ
Reactivity: Mouse b haplotype;Cross-reacts with H-2Dd MHC class I alloantigen. Does not react with other haplotypes (e.g.;f;k;p;q;r;s)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR