Pipette Box, White PVC, Adjustable, TrippNT
Supplier: TrippNT
50492
11376-228EA 248.64 USD
11376-228 11376-234
Pipette Box, White PVC, Adjustable, TrippNT
Trays/Dishes/Pots Pipette Trays
The inside distance of this storage box is 25" making it perfect for long, large volume, volumetric pipets.
  • Stores and separates long pipettes
  • Dividers can move to accommodate storage needs
  • Choose from 2 sizes
  • White PVC construction with optional acrylic lid
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement Lifetime Guarantee
The hinged lid (which is optional) protects supplies from dust or dirt and extra protection from breakage. You can also change the size of the compartments with the adjustable dividers so it fits exactly the number and size of your items.