Packer Bottles, Amber, Wide Mouth
Supplier: Qorpak
GLC-02197 GLC-05372 GLC-07129 GLC-07132 GLA-00932 GLA-00921 GLC-02127 GLC-05972 GLC-05971 GLC-02194 GLC-05968 GLC-05970 GLC-05969 GLA-00910 GLA-00929 GLC-05933 GLC-05926 GLC-05942 GLC-05924 GLC-05941 GLC-06063 GLA-05149 GLA-00911 GLA-00917 GLA-00922 GLA-00912 GLA-00913 GLC-02089 GLC-02088 GLC-02106 GLC-02105 GLC-02203 GLC-02202 GLC-02125 GLC-02124 GLC-02174 GLC-02081 GLC-02096 GLC-02190 GLC-02118 GLC-02135 GLC-02161 GLC-02085 GLC-02101 GLC-02196 GLC-02142 GLC-02169 GLA-00930 GLA-00914 GLA-00926 GLA-00923 GLA-00919 GLC-02097 GLC-02191 GLC-02082 GLC-02114 GLC-09083 GLC-02184 GLC-06609 GLC-02132 GLC-02183 GLC-07130 GLC-07131 GLC-02092 239211 272169 272168 272167 272166 272165 272164 272163 272162 272161 272160 272156 GLC-02077 GLC-02109 GLC-02128 GLC-02153 GLC-06733 GLA-00927 GLA-06579
11311-204CS 556.86 USD
11311-204 11311-528 11311-530 11311-532 11311-534 11311-540 11311-584 11311-586 11311-588 16054-156 16054-158 16054-400 16054-402 16054-404 16054-416 16054-418 16054-420 16054-428 16054-430 16054-432 16054-434 16054-436 16054-438 16054-440 16054-442 16054-444 16087-014 16087-032 16087-034 16087-036 16151-596 16151-608 16179-988 16179-999 16180-003 16180-014 16180-025 16180-036 16180-047 16180-058 16180-091 16180-095 16180-207 16180-229 16180-240 16180-262 16180-284 16180-309 16180-444 16180-488 16180-681 16180-706 16180-728 16180-852 16180-874 16180-879 16218-174 16218-226 16218-228 16218-230 36319-156 36319-158 36319-250 36319-263 36319-265 36319-266 36319-268 71000-110 71000-112 71000-114 71000-116 71000-118 71000-120 89140-224 93001-568 89204-848 89027-040 89027-042 10027-440 10027-444 10027-450 10158-494 10158-496 75993-598 76170-332 76170-334 76170-336 76170-338 76170-340 76170-342 76170-344 76170-346 76419-744 76419-746 76419-748 76419-750 76419-752 76419-754 76419-756 76419-758 76419-760 76419-762 76419-766 76419-768 76419-770 76487-192 76487-194 76487-196 76487-198 76487-200 76539-350 76539-354
Packer Bottles, Amber, Wide Mouth
Bottles Glass Bottles
Amber wide mouth Packer bottles feature a wide opening making them the ideal general-purpose bottle for solids.
  • Packer bottles are made of glass
  • Ideal for light sensitive products
Applications include pharmaceutical, chemical, and environmental.
Ordering information: Bottles 11311-528, 11311-530, 11311-532, 11311-534, 11311-540, 10158-494 and 10158-496 are supplied Ultra Cleaned. Bottles 36319-156, 36319-158, 36319-250, 36319-253, 36319-256, 36319-265, 36319-268, and are supplied pre-cleaned for volatiles. Bottles 89204-848, BA11311-362CRC, 76487-192, 76487-194, 76487-196, 76487-198 and 76487-200 are supplied pre-cleaned and certified for semi-volatiles.