Density/Specific Gravity Kits, METTLER TOLEDO®
Supplier: Mettler Toledo
210260
11297-240EA 1234.35 USD
11297-240 62410-628
Density/Specific Gravity Kits, METTLER TOLEDO®
Density Meters
This kit works with balance to determine the density and specific gravity of liquids.
  • Convenient and time-saving kit
  • Based on buoyancy method (Archimedes' principle)