Ion Selective Half Cell Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
51340300 51340400 51340500 51340600 51340800 51340263 51340900 51340700 51341000
11277-330EA 1450.63 USD
11277-330 11277-332 11277-334 11277-336 11277-340 11277-350 11277-342 11277-338 11277-344
Ion Selective Half Cell Electrode, METTLER TOLEDO®
Electrodes ISE Electrodes
Ion Selective half-cells electrodes consist of two elements, a universal shaft and an ion-specific membrane module.
  • Electrolyte filled
  • Ready to use
  • Tested for wet chemistry
  • Passes stringent requirements
  • Fast response time and requires no maintenance
This module may be exchanged to measure desired ions.
Each ion-selective electrode (ISE) and each membrane module has been tested for wet-chemistry applications. It has to pass the stringent requirements of several direct measurements as well as a typical titration application.
The sodium electrode has a similar construction to a normal pH electrode but uses membrane glass with a higher selectivity for sodium.
Ordering information: Membrane kits available should be ordered separately.
Delivery information: The membrane kit comes with an identification ring (ID ring) and an adapter sleeve which will fit any METTLER TOLEDO titration stand.