Redox Buffer Solution
Supplier: Mettler Toledo
51350060
11238-454EA
60.45
USD
11238-454
Redox Buffer Solution
Buffers
pH Calibration Buffers
Redox buffer solutions are used for verification of all common redox sensors.
- Temperature dependent
- Used in quality assurance systems
A table on every bottle label indicates redox values at different temperatures and hence not used for calibration purposes.
Delivery information: Supplied with a quality inspection certificate.