LABRACK™ Rack for VWR® Sample Bags
Supplier: VWR International
SA4-7012-VW
11216-910EA 246.43 USD
11216-910
LABRACK™ Rack for VWR® Sample Bags
Racks Bag Racks
This stainless steel rack allows you to carry multiple bags easily and safely. It can go in the incubator and autoclave, and can be stacked for space saving.
  • Temperature resistant from −80 °C to +80 °C
  • Autoclavable
  • Made of 100% stainless steel
  • Can store up to ten blender bags
Ideal for sample batch preparation prior to blending as well as sample storage after blending.