Mr.Tuff Pail with Lid, High Density Polyethylene, Nasco
Supplier: Nasco
C05575N
11216-015EA 26.93 USD
11216-015
Mr.Tuff Pail with Lid, High Density Polyethylene, Nasco
Buckets
A sturdy, white, top-quality pail for all uses, made of high-density polyethylene with a leakproof lid.
  • Easy to clean and can be steam sterilized
  • Rust-, dent-, and crack-free
  • Galvanized wire bail with plastic grip
  • Withstand corrosive or noncorrosive products
  • Extremely durable, resilient, and has seamless construction
Certifications: Complies with FDA specifications.