SP Bel-Art Dialysis Bag Clip Holders, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18237-0000
11215-308BG 47.86 USD
11215-308
SP Bel-Art Dialysis Bag Clip Holders, Bel-Art Products, a part of SP
Racks Bag Racks
Suspend dialysis bags in large beakers or pitchers up to 0.63 cm (1/4") thick with these strong clips.
  • Smooth polypropylene construction
  • Autoclavable
  • Easy to mark with sequence numbers
Bags slide easily into the slot and then the holder can be clipped onto the container’s edge. Smooth polypropylene surface may be marked with identifying sequence numbers.