Terminal® Broken Glass Benchtop Keeper™ Container, Whitney Products
Supplier: Heathrow Scientific
BH2004
11214-713CS 324.35 USD
11214-713
Terminal® Broken Glass Benchtop Keeper™ Container, Whitney Products
Boxes Waste Disposal Boxes
For the safe collection of non-biohazardous broken glass
  • Contents are clearly identified
  • Plastic-coated lining for leak resistance
  • Self-locking container is autoclavable
Helps protect against accidental injuries.