Kapak Autoclavable FoilPAK Pouches, Ampac Flexibles
Supplier: Ampac Flexibles
KSP-150-1MB
11213-852CS 2970.71 USD
11213-852
Kapak Autoclavable FoilPAK Pouches, Ampac Flexibles
Bags Sterilization Pouches
Ampac's autoclaveable FoilPAK (KSP-150) is a 3ply structure consisting of polyester, foil, and cast polypropylene (PET/Foil/CPP).
This pouch can withstand temperatures up to 130 °C (266 °F).