Hydrochloric acid

Supplier: SEASTAR CHEMICALS

Synonyms: Hydrochloric acid solution, Muriatic acid

S010401-ANNQ09
11020-892EA 355.54 USD
11020-892
Hydrochloric acid
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR