Silica sand 70 Mesh

Danger

16153374
11019-851EA 62.84 USD
11019-851
Silica sand 70 Mesh
Silica
Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112945-52-5
EINECS: 238-878-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR