STARRBIDE Crucibles, Morgan Advanced Materials
Supplier: MORGAN ADVANCED MATERIALS
670-7000-0005186
11019-830EA 2495.08 USD
11019-830
STARRBIDE Crucibles, Morgan Advanced Materials
Crucibles
STARRBIDE is a high-quality, tar-bonded silicon carbide material designed to provide reliable performance across the full range of non-ferrous melting and holding applications.
  • Good thermal shock resistance
  • Good resistance to attack by chemical treatment agents
  • High mechanical strength
STARRBIDE crucibles are colored black. They are available in a comprehensive range of shapes and sizes to suit most end-user requirements.
Certifications: Manufactured to ISO9001:2008 quality standards.