HIMELT Crucibles, Morgan Advanced Materials
Supplier: MORGAN ADVANCED MATERIALS
694-4000-0002146
11019-828EA 2418.78 USD
11019-828
HIMELT Crucibles, Morgan Advanced Materials
Crucibles
HIMELT high-quality, carbon-bonded silicon carbide crucibles are manufactured using the latest roller-forming techniques and are designed to cater to a range of non-ferrous melting applications.
  • Fast melting speed through high consistent thermal conductivity
  • Excellent thermal shock resistance
  • High resistance to oxidation
  • Good erosion resistance
  • Good resistance to corrosive attack by chemical treatment agents
HIMELT crucibles are colored bright red. They are available in a comprehensive range of shapes and sizes to suit most end user requirements.
Certifications: Manufactured from premium grade raw materials under an ISO 9001:2008 quality management system.