Riffle Pan, New Liskeard Sheet Metal
Supplier: NEW LISKEARD SHEET METAL WORKS
SP111
11019-604EA 45.35 USD
11019-604
Riffle Pan, New Liskeard Sheet Metal
Samplers Solid Samplers
Pan is designed to facilitate rapid and even splitting of solid samples and other dry substances.
Pan dimensions are 12 x 5 x 3.5".