Kaowool™ Refractory Fiber Blanket, Morgan Advanced Materials
Supplier: DISTRIBUTION INTERNATIONAL
CFBT8102CBT
11019-480EA 293.91 USD
11019-480
Kaowool™ Refractory Fiber Blanket, Morgan Advanced Materials
Insulation
Kaowool™ Blankets are made from bulk fibers produced by the most modern spinning process.
  • Excellent thermal shock resistance
  • No smoke emission due to binder burn out
  • Low heat storage
  • Good acoustic properties
  • Easy to install
  • Unaffected by most chemicals, except hydrofluoric acid, phosphoric acids, and strong alkali
Applications include furnace linings (all industries: iron & steel, non-ferrous, aluminium, bricks and ceramics/pottery), petrochemical (crackers, heaters, and ducts), cladding/lining waste-heat ducts, exhaust cladding, acoustic insulation, steam turbine insulation, hot boxes, and domestic appliances.
This produces fibers of exceptional length and uniform distribution, which are ideal for manufacturing blankets of controlled density, high tensile strength, and resiliency.