Lead, foil

Supplier: ALPHA ASSEMBLY SOLUTIONS, INC.
Danger

142733
11019-412EA 190.49 USD
11019-412
Lead, foil
Lead

Lead Foil is typically used in both Fire Assay bullion work and cupellation.  For example to wrap bullion samples, or for additional cupellation purposes.

Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now

Specification Test Results

Thickness 0.1 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR