Deuterium Arc Lamp. Model P706, Photron
Supplier: PHOTRON PTY LTD.
P706
11018-984EA 593.12 USD
11018-984
Deuterium Arc Lamp. Model P706, Photron
Lights Deuterium Lamps
UV light source for spectroscopy
Caution: Care must be taken not to contact lamp with bare hand, temperature can reach over 200°C during operation. UV Radiation is harmful to the eyes. Do not observe light output directly. Deuterium lamps operate at high voltages. Extreme caution is required to prevent electrical shock.
For use with Varian instruments. 3-pin housing terminal.