Hollow Cathode Lamps to Suit PerkinElmer Systems, Photron
Supplier: PHOTRON PTY LTD.
P914 P926 P921 P933 P951
CA11018-216EA 540.3 USD
CA11018-216 CA11018-240 11018-230 11018-254 11018-290
Hollow Cathode Lamps to Suit PerkinElmer Systems, Photron
Lights Hollow Cathode Lamps
Photron's P900 (51mm or 2.0” Diameter) series is designed for direct use in AAS systems manufactured by PerkinElmer.
These hollow cathode lamps are produced to provide direct use without adapters in all PerkinElmer AAS instruments. All windows on these lamps are fully fused and contain no "gassy" adhesives which drastically reduce shelf life. Totally hermetic glass seals ensures a clean fill gas and cathode for the life of the lamp and most importantly a shelf life longer than 5 years. The application of the same glass cathode shielding technique used in all Photron hollow cathode lamps gives maximum stability and reliability throughout lamp life. Due to the elegantly simple design of the electrode geometry of Photron lamps, less mass is present within the lamp, reducing outgassing and making the lamp more resistant to breakage from lateral shock.
Ordering information: Contact your Avantor sales representative for more information. Wavelength listed is the primary wavelength.