Crucible Tongs, Steel, Nickel, Webber's Welding
Supplier: Webber's Welding
FA115A
CA11017-654EA 674.79 USD
CA11017-654 11017-654
Crucible Tongs, Steel, Nickel, Webber's Welding
Tongs
These crucible tongs are made of mild steel for durable and dependable service.
Their longer length increases user safety, by distancing the user from the oven or furnace.