Whatman™ Grade 591 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
10311387
11008-572PK 1417.44 USD
11008-572
Whatman™ Grade 591 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
These cellulose filters are used in qualitative analytical techniques to determine and identify materials.
  • Thick filter paper with very high loading capacity
  • High absorbency and increased wet strength
A standard grade filter for fast filtration of medium to coarse precipitates.