Nunclon™ Delta MultiDish Inserts, Thermo Scientific
Supplier: Thermo Fisher Scientific
140627 140644 140654 140660 140640 140642 140663 140668 140620 140629 140656 140652
89096-058CS 438.14 USD
89096-058 89094-370 89177-136 89177-140 89094-366 89094-368 89177-142 89177-144 89177-132 89177-134 89177-138 11006-424
Nunclon™ Delta MultiDish Inserts, Thermo Scientific
Culture Dishes Cell Culture Dishes
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes
Use for transport studies, toxicity tests, chemotaxis studies, and electron microscopy. They are also suitable for phase contrast and Nomarski optic systems. Inserts have been treated and tested for cell attachment, providing easy cultivation of most cell types without the need for matrix coating. They feature tabs for better handling. Nontoxic. Nonpyrogenic.
Ordering information: Packed in resealable bags.