Overhead Arms, Production Basics
Supplier: Production Basics
8538
11005-382EA 202.83 USD
11005-382
Overhead Arms, Production Basics
Furniture Casework
Use with our Tool Trolley Systems or for applications requiring air, power or light to be positioned above the operator.
  • Arms extend up to 35.56 cm (14") above the top of the Uprights or Frame
  • 25 lbs. load capacity
For use with on any C-Leg Lab Bench. Add uprights to mount accessories on RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench, Basic Organizer or Easy Lift 4-post Lab Bench
Ordering information: Standard accessory color is Black. Other standard frame color (Light Neutral/Almond, Blue, White, or Grey) available by request. Accessory is ready to mount on a C-Leg Lab Bench; or otherwise must purchase Uprights, sold separately, if mounting on a RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench or Easy Lift 4-post Lab Bench.
Delivery information: Includes Overhead Arms (1), Pair, Parts and Hardware Packet (1).