Autokit 3-HB R1 Set, FUJIFILM Irvine Scientific
Supplier: Wako Life Sciences
417-73501
11001-664EA 1863.86 USD
11001-664
Autokit 3-HB R1 Set, FUJIFILM Irvine Scientific
Assays Protein Assays
The Autokit 3-Hb R1 Set is an in vitro assay for the quantitative determination of 3-hydroxybutyrate (3-HB) in serum or plasma.
  • Utilizes cyclic enzymatic reactions
  • High sensitivity and high specificity
  • Quantitative determination of 3-HB in serum or plasma
The Autokit 3-Hb R1 Set is an in vitro assay for the quantitative determination of 3-hydroxybutyrate (3-HB) in serum or plasma. It utilizes cyclic enzymatic reactions for high sensitivity and high specificity.